Order Entry
Austria
ContactUsLinkComponent
Carbon disulphide ≥99.9% (by GC, corrected for water content), ULTRA RESI-ANALYZED® for spectroscopy, J.T.Baker®
Flammable
Health
Irritant
Carbon disulphide ≥99.9% (by GC, corrected for water content), ULTRA RESI-ANALYZED® for spectroscopy, J.T.Baker®
Catalog # E350-01
Supplier:  Avantor
CAS Number:  
undefined
Flammable
Health
Irritant
Carbon disulphide ≥99.9% (by GC, corrected for water content), ULTRA RESI-ANALYZED® for spectroscopy, J.T.Baker®
Catalog # E350-01
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, clear, PVC-coated
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • ACS - Assay (CS₂) (by GC, corrected for water):
    ≥99.9 %
  • ACS - Trace Organic Impurities (as benzene) (by GC):
    ≤1 ppm
  • ACS - Color (APHA):
    ≤10
  • ACS - Residue after Evaporation:
    ≤15 ppm
  • ACS - Hydrogen Sulfide (H₂S):
    ≤1.5 ppm
  • ACS - Sulfur Dioxide (SO₂):
    ≤2.5 ppm
  • ACS - Water (H₂O)(by Karl Fischer titrn):
    ≤0.05 %
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 750 nm:
    ≤0.01
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 500 nm:
    ≤0.01
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 410 nm:
    ≤0.05
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 403 nm:
    ≤0.10
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 394 nm:
    ≤0.25
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 388 nm:
    ≤0.50
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 385 nm:
    ≤1.00
  • Boiling Point (typical)................. 46.3 °C.:
    2.68 - 3.34, 2.23 - 2.68, 0.75 - 2.19
  • Windows of Near Infrared Transmittance (10-mm path vs. air 80-90% T),:
    2.5 - 4.3, 4.8 - 6.2, 7.1 - 16.0

Specification Test Results

About this item

 :
 :76.14 g/mol
Merck Index:13,01821
Boiling Pt:46.2 °C (1013 hPa)
Melting Pt:–111.6 °C
Density:1.2632 g/cm³ (20 °C)
Flash Pt:–30 °C
EINECS:200-843-6
UN:1131
ADR:3 (6.1),I
 :GHS02
 :GHS08
 :GHS07
Storage Temperature:Ambient
CAS Number:75-15-0
Formula:CS₂
MDL Number:MFCD00011321