Trident direct guard cartridge system
Supplier: Restek
Certificates
About this item
Trident direct system can be used to select the right level of protection for analysis. It provides the protection against particulate matter and moderate protection against irreversibly adsorbed compounds.
- Easy to use
- Low dead volume
- Leak-free cartridge
The system is available in configurations to match different protection levels such as in-line filter, in-line filter with holder for 10 mm guard cartridge. The guard cartridges are available in 2,1 and 4,0 mm ID and are interchangeable within the appropriate length holder.
Frequently Bought Together