Order Entry
Austria
ContactUsLinkComponent
Sulphuric acid 0.13 mol/l (0.26 N) in aqueous solution, AVS TITRINORM
Sulphuric acid 0.13 mol/l (0.26 N) in aqueous solution, AVS TITRINORM
Catalog # 86103.360
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sulphuric acid 0.13 mol/l (0.26 N) in aqueous solution, AVS TITRINORM
Catalog # 86103.360
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    5 L
  • Seq:
    5190

Specifications

Specification Test Results

  • Titer:
    0.1250 --> 0.1350 mol/l

Specification Test Results

About this item

 :
 :98.08 g/mol
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589