1,1,1,3,3,3-Hexafluoro-2-propanol ≥99% (by GC, corrected for water content), BAKER ANALYZED®, J.T.Baker®
Catalog # N151-05
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle, amber, narrow mouth
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Assay (C₃H₂F₆0) (by GC, corrected for water):≥99 %
- Color (APHA):≤10
- Residue after Evaporation:≤0.100 %
- Water (H₂O)(by Karl Fischer titrn):≤0.05 %
Specification Test Results
About this item
:
Merck Index:13,09598
Boiling Pt:59.1 °C (1013 hPa)
Melting Pt:–96 °C
Density:1.492 g/cm³ (20 °C)
Flash Pt:>100 °C
Storage Temperature:Ambient
EINECS:213-059-4
UN:1760
ADR:8,II
:168.04 g/mol
:GHS05
:GHS07
CAS Number:920-66-1
Formula:C₃H₂F₆O
MDL Number:MFCD00011651