Order Entry
Austria
ContactUsLinkComponent
 
Dimethyl sulphoxide ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
    
 
undefined
Dimethyl sulphoxide ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
    
 

 

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default

 

 

  • Assay (GC):
    Min. 99.70 %
  • Non-volatile matter:
    Max. 0.002 %
  • Water (Karl Fischer):
    Max. 0.2 %
  • Absorbance at 350 nm:
    Max. 0.01
  • Absorbance at 300 nm:
    Max. 0.10
  • Absorbance 280 nm:
    Max. 0.30
  • Absorbance at 270 nm:
    Max. 0.70
  • Vapor Pressure:
    0.42 mmHg ( 20 °C)
  • Explosion Limit:
    42 %, 63 °F
  • Autoignition Temperature:
    573 °F
  • Vapor Density:
    2.7 (vs air)

 

 

Frequently Bought Together