Replacement cap frit filters for Trident guard cartridges
Supplier: Restek
Certificates
About this item
The removable cap frit filters in a Trident help to prevent clogged cartridges and to extend the life of the column and cartridge.
Frequently Bought Together