Oxidase sticks for microbiology
APPC416444.2326
:
- Pk:50
- Description:Oxidase sticks for microbiology
Plastic sticks used for the easy and rapid detection of the cytochrome oxidase enzyme in microbiological diagnosis. Oxidase sticks have a much higher stability compared to the traditional laboratory test, because the tetramethyl-p-phenylenediamine reagent is attached to the pad.
: *The product(s) on this page are outside the scope of the EU Directive 2019/904