Order Entry
Austria
ContactUsLinkComponent
 
Analytical standard, Benthiavalicarb-isopropyl, reference material, Honeywell Fluka™
  HONCR1365-10MG
 :  
Analytical standard, Benthiavalicarb-isopropyl, reference material, Honeywell Fluka™
  HONCR1365-10MG
 :  

 

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    177406-68-7
  • Description:
    Benthiavalicarb-isopropyl
  • Melting point:
    168…173 °C
  • Formula:
    C₁₈H₂₄FN₃O₃S
  • Storage temperature:
    Ambient
  • Description:
    Benthiavalicarb-isopropyl

 

 

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results