Analytical standard, Desogestrel, reference material, Honeywell Fluka™
HONCR1358-25MG
:
- Pack type:Glass Bottle
- Pk:25 mg
- Standards:Analytical standard, neat
- CAS No.:54024-22-5
- Description:Desogestrel
- Environmentally Preferable:
- Melting point:105…110 °C
- Formula:C₂₂H₃₀O
- Storage temperature:Ambient
- Description:Desogestrel
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results