Analytical standard, Tetraconazole, reference material, Honeywell Fluka™
Artikelnr. HONCR1305-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:100 mg
- Standards:Analytical standard, neat
- CAS No.:112281-77-3
- Description:Tetraconazole
- Formula:C₁₃H₁₁Cl₂F₄N₃O
- Storage temperature:Ambient
- Description:Tetraconazole
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results