Order Entry
Austria
ContactUsLinkComponent
 
Analytical standard, Prochloraz Metabolite BTS44595, reference material, Honeywell Fluka™
Catalog # HONCR1274-10MG
CAS Number:  
Analytical standard, Prochloraz Metabolite BTS44595, reference material, Honeywell Fluka™
Catalog # HONCR1274-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    139520-94-8
  • Description:
    Prochloraz Metabolite BTS44595
  • Melting point:
    54…56 °C
  • Formula:
    C₁₂H₁₅Cl₃N₂O₂
  • Storage temperature:
    Ambient
  • Description:
    Prochloraz Metabolite BTS44595

Specifications

About this item

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results