Analytical standard, Flumequine-(1,2,carboxy-13C3), reference material, Honeywell Fluka™
Catalog # HONCR1233-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:1185049-09-5
- Description:Flumequine-(1,2,carboxy-13C3)
- Environmentally Preferable:
- Melting point:254 °C
- Formula:¹³C₃C₁₁H₁₂FNO₃
- Storage temperature:Ambient
- Description:Flumequine-(1,2,carboxy-13C3)
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results