Order Entry
Austria
ContactUsLinkComponent
 
Analytical standard, Carbuterol acetate, reference material, Honeywell Fluka™
  HONCR1231-10MG
 :  
Analytical standard, Carbuterol acetate, reference material, Honeywell Fluka™
  HONCR1231-10MG
 :  

 

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    1613439-57-8
  • Description:
    Carbuterol acetate
  • Melting point:
    110 °C
  • Formula:
    C₁₃H₂₁N₃O₃ · C₂H₄O₂
  • Storage temperature:
    2…8 °C
  • Description:
    Carbuterol acetate

 

 

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results