Analytical standard, Uniconazole, reference material, Honeywell Fluka™
HONCR1183-50MG
:
- Pack type:Glass Bottle
- Pk:50 mg
- Standards:Analytical standard, neat
- CAS No.:83657-22-1
- Description:Uniconazole
- Environmentally Preferable:
- Melting point:147…164 °C
- Formula:C₁₅H₁₈ClN₃O
- Storage temperature:Ambient
- Description:Uniconazole
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results