Analytical standard, Pyrazolynate, reference material, Honeywell Fluka™
Catalog # HONCR1101-25MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:25 mg
- Standards:Analytical standard, neat
- CAS No.:58011-68-0
- Description:Pyrazolynate
- Melting point:119 °C
- Formula:C₁₉H₁₆Cl₂N₂O₄S
- Storage temperature:2…8 °C
- Description:Pyrazolynate
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results