Order Entry
Austria
ContactUsLinkComponent
 
Buffer capsules, coloured coded
Catalog # REGECP1100
Supplier:  REAGECON
CAS Number:  
Buffer capsules, coloured coded
Catalog # REGECP1100
Supplier:  REAGECON
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, amber
  • Pk:
    50
  • Reference temperature:
    25 °C
  • Colour:
    Blue
  • Description:
    pH buffer capsules, reference standard
  • Seq:
    04
  • pH accuracy:
    ±0,02
  • pH value:
    10,00

Specifications

About this item

Certifications: Certified by Reagecon’s ISO 17025 accredited test method. NIST traceable.