Culture media, in petri dishes, tryptic soy agar, with LTHThio
Supplier: MERCK LIFE SCIENCE
Certificates
About this item
TSA can be used in environmental monitoring for isolators and clean rooms (surface and air monitoring). Also used for the detection of fungi and bacteria.
TSA contains CASO agar, casein soybean digest agar with lecithin, tween, histidin and thiosulphate, tryptone soya agar, tryptic soy agar with neutraliser.
Packaging: Packaged in fibre case.
Specifications
- Storage conditions:15...25 °C