Order Entry
Austria
ContactUsLinkComponent
di-Sodium tetraborate decahydrate 99.0-105.0%, GenAR® NF for biotechnology, Macron Fine Chemicals™
 
 
di-Sodium tetraborate decahydrate 99.0-105.0%, GenAR® NF for biotechnology, Macron Fine Chemicals™
  7792-88
 :  
undefined
 
 
di-Sodium tetraborate decahydrate 99.0-105.0%, GenAR® NF for biotechnology, Macron Fine Chemicals™
  7792-88
 :  

 

  • Pack type:
    PE pail, screw top
  • Pk:
    12 kg
  • Seq:
    4187

 

 

  • Preserve in Tight Containers:
    99.0 - 105.0%
  • CAUTION: For Manufacturing, processing or repackaging:
    Passes Test
  • Bulk Pharmaceutical Chemical:
    Passes Test
  • NF - Assay (Na₂B₄O₇·10H₂O):
    99.0 - 105.0 %
  • NF - Carbonate and Bicarbonate:
    Passes Test
  • NF - Identification A:
    Passes Test
  • NF - Identification B:
    Passes Test
  • EP/BP - Assay (Na₂B₄O₇·10H₂O):
    99.0 - 103.0 %
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Identification C:
    Passes Test
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - pH of 4% Solution at 25°C:
    9.0 - 9.6
  • EP/BP - Sulfate (SO₄):
    ≤50 ppm
  • EP/BP - Ammonium (NH₄):
    ≤10 ppm
  • EP/BP - Arsenic (As):
    ≤5 ppm
  • EP/BP - Calcium (Ca):
    ≤100 ppm
  • JP - Assay (Na₂B₄O₇·10H₂O):
    99.0 - 103.0 %
  • JP - Arsenic (As):
    ≤5 ppm
  • JP - Identification A:
    Passes Test
  • JP - Identification B:
    Passes Test
  • JP - pH (1 in 20):
    9.1 - 9.6
  • JP - Clarity and Color of Solution:
    Passes Test
  • JP - Carbonate and Bicarbonate:
    Passes Test
  • JP - Heavy Metals (as Pb):
    ≤20 ppm

 

 

 :
 :381.37 g/mol
 :13,08662
 :320 °C (1013 hPa)
 :75 °C
 :1.73 g/cm³
 :215-540-4
 :01-2119490790-32
 :GHS07
 :GHS08
 :Ambient
 :1303-96-4
 :Na₂B₄O₇·10H₂O
 :MFCD00149193