Flash chromatography cartridges, filter elements, CHROMABOND® Flash DL
Supplier: MACHEREY-NAGEL
Certificates
About this item
Frequently Bought Together
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Magnetic Stirring Bars, Cross-Shaped
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Flash chomatography cartridges, empty colums, CHROMABOND® Flash DL and connecting kits
Supplier: MACHEREY-NAGEL
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Exhaust filters, V3.0
Supplier: S.C.A.T.
N-Ethyldiisopropylamine ≥99.5% for peptide synthesis
Supplier: VWR Chemicals
VWR®, Screw Neck Headspace Vials ND18
Supplier: VWR Collection
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals