ContactUsLinkComponent
Cleanroom shoe cover, PP/CPE, BioClean Dual™
Cleanroom shoe cover, PP/CPE, BioClean Dual™
Catalog # NITRS-BDOS
Supplier:  Nitritex
undefined
Cleanroom shoe cover, PP/CPE, BioClean Dual™
Catalog # NITRS-BDOS
Supplier:  Nitritex
Specifications
  • Sterile:
    +
  • Garment application:
    Cleanroom
  • Colour:
    White
  • Length:
    400 mm
  • Pk:
    150 PAIR
  • Cleanroom class:
    10/ISO 4
Specifications
About this item
Disposable shoe covers made from BioClean Dual™, a low linting spunbonded non-woven polypropylene fabric with embossed cast polyethylene sole for a secure footing.
  • Durable
  • Heavy duty slip-resistant sole
  • Latex-free elastic
Suitable for use in ISO Class 4 environment.
Certifications: Manufactured in a facility holding ISO 9001:2008 certification and processed in an NEBB certified ISO Class 4 environment.
Packaging: One pair per sealed inner PE bag, 15 inner bags per sealed outer bag, 10 outer bags per lined carton (150 pairs).