ContactUsLinkComponent
Battery powered engraving tool, SP Bel-Art
Battery powered engraving tool, SP Bel-Art
Catalog # BELAF441500005
undefined
Battery powered engraving tool, SP Bel-Art
Catalog # BELAF441500005
Specifications
  • Description:
    Battery powered engraving tool
  • Pk:
    1
Specifications
About this item
Battery powered engraving tool to permanently mark most materials including glass, plastic, wood, metal, bone, and stone.
  • Cordless battery power makes it portable for use virtually anywhere indoors or out in the field
  • Marks will not fade if exposed to chemicals or sunlight
  • Operates on two 'AA' batteries
Simply hold the engraving tool like a pen, press the power button, apply light pressure and write on test tubes, bottles, racks, keys, equipment, tools, electronics and more.
Ordering information: Convenient polyethylene stand (sold separately) with safe place to keep extra tip (tip included). AA batteries are not included.