ContactUsLinkComponent
Disposable particle filtering respirators, P391
Disposable particle filtering respirators, P391
Catalog # PRTWP391WHR
Supplier:  Portwest
undefined
Disposable particle filtering respirators, P391
Catalog # PRTWP391WHR
Supplier:  Portwest
Specifications
  • Type:
    P391
  • Pk:
    10
  • With valve:
    +
  • Class:
    FFP3
Specifications
About this item
Eagle FFP3 folding white mask with seven dynamic parts provides excellent facial movement with maximum comfort. Valved and with inner foam to increase the comfort of the wearer.
  • High performance exhalation valve to reduce heat and provide comfort when used in hot and humid conditions
  • Large inner space to help the user to breathe more easily
  • Flexible shape which allows the mask to be folded flat
Dolomite optional clogging test (D) to provide more comfortable breathing and longer lasting performance of filtration.
Certifications: EN 149 FFP3 (NR D)
Packaging: Individually wrapped and packed inside hygienic polybag for vending machines.