ContactUsLinkComponent
USB interface for Testo 174T and 174H
USB interface for Testo 174T and 174H
Catalog # 620-1838
Supplier:  Testo
undefined
USB interface for Testo 174T and 174H
Catalog # 620-1838
Supplier:  Testo
Specifications
  • Description:
    For programming and readout of loggers testo 174T and testo 174H
  • Seq:
    44
  • Pk:
    1
Specifications
About this item
USB interface for programming and readout of the loggers testo 174T and testo 174H.