Order Entry
Austria
ContactUsLinkComponent
TraceCERT®, Supelco® Chromate Standard for IC, CRM
TraceCERT®, Supelco® Chromate Standard for IC, CRM
Catalog # SUPL40121-100ML
Supplier:  Merck
CAS Number:  
TraceCERT®, Supelco® Chromate Standard for IC, CRM
Catalog # SUPL40121-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg/l
  • Matrix:
    H₂O
  • Pk:
    100 mL
  • Description:
    Chromate standard

Specifications

About this item

Single component solution suitable as ion chromatography standard.

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34).