Order Entry
Austria
ContactUsLinkComponent
EDTA disodium salt dihydrate 99.0-101.0% (by complexometric analysis) USP, BP, Ph. Eur., pharma grade
Irritant
Health
EDTA disodium salt dihydrate 99.0-101.0% (by complexometric analysis) USP, BP, Ph. Eur., pharma grade
Supplier:  PanReac AppliChem
Certificates
undefined
Irritant
Health
EDTA disodium salt dihydrate 99.0-101.0% (by complexometric analysis) USP, BP, Ph. Eur., pharma grade
Supplier:  PanReac AppliChem
Certificates

Specification Test Results

  • Assay (Complexometric):
    99.0 - 101.0%
  • Identity According to Pharmacopoeias:
    Passes Test
  • pH of 5% Solution:
    4.0 - 5.0
  • Insoluble Matter in H₂O:
    0.01%
  • Loss on Drying at 150 °C (6 Hours):
    8.7 - 11.4%
  • Chloride (Cl):
    0.02%
  • Sulphate (SO₄):
    0.1%
  • Residual Solvents (Ph. Eur./USP):
    Passes Test
  • Nitrilotriacetic Acid [(CH₂COOH)₃N]:
    0.1%
  • Heavy Metals (as Pb):
    0.001%
  • Ca:
    Passes Test
  • Fe:
    0.0005%

Specification Test Results

Related Information

Frequently Bought Together