Order Entry
Austria
ContactUsLinkComponent
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) substrate ≥99%, Molecular Biology Grade
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) substrate ≥99%, Molecular Biology Grade
Catalog # OZBIXG31000
Supplier:  OZ BIOSCIENCES
CAS Number:  
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) substrate ≥99%, Molecular Biology Grade
Catalog # OZBIXG31000
Supplier:  OZ BIOSCIENCES
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :408.63 g/mol
CAS Number:7240-90-6
Formula:C₁₄H₁₅BrClNO₆
MDL Number:MFCD00005666