Transfection reagent, in vivo PolyMag™
In vivo PolyMag™ transfection reagent has been designed for in vivo targeted transfection of various types of nucleic acids, such as DNA, RNA, oligonucleotides, using magnetic nanoparticles coated with specific polymers. DNA complexes can be easily administrated through various injection routes, such as systemic administration (intravenous, intra-artery) or local administration (intratumoral, intracerebroventricular).
- Increased transfection efficiency
- Targeted process (magnetically-driven)
- Reduction of the systemic dissemination of vectors during injection
- Reduction of the vector doses
- Work under non permissive conditions (hypothermia, physiological flow conditions)
- Penetration of the vector into tissues
- MInimised toxicity
- Universal suitable for all nucleic acids
Store at +4 °C.
: Needs to be used with a magnetic plate, available as a separate accessory or as part of a starter kit.
: Shipped at room temperature.
- DNA:Yes
- in vivo:Yes
- mRNA:Yes
- Oligos:Yes
- shRNA:Yes
- siRNA:Yes
Frequently Bought Together