Orders LinkContactUsLinkComponent
Sample loop kit
Sample loop kit
Assorted sample loops, PTFE (25, 50, 100, 200, 500 µl)
Catalog # 18-0404-01
Supplier:   Cytiva
Sample loop kit
Catalog # 18-0404-01
Supplier:  Cytiva
Sample loop kit
Sample loop kit
Assorted sample loops, PTFE (25, 50, 100, 200, 500 µl)
Price
525,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
Each sample loop consists of tubing 1,8 mm OD and two tubing connectors (M6).
Delivery information: The kit contains: 1×25 µl sample loop 0,5 mm ID; 1×50 µl sample loop 0,5 mm ID; 1×100 µl sample loop 0,5 mm ID; 1×200 µl sample loop 0,8 mm ID; 1×500 µl sample loop 0,8 mm ID.
Specifications
  • Description:
    Assorted sample loops, PTFE (25, 50, 100, 200, 500 µl)