Quick-release clamps for flat flange connections
Supplier: Lenz Laborglas
Jump to:
About this item
Stainless steel, rust-proof.
- Simple assembly and disassembly
- With three holding segments
- Quick release
For clamping reaction vessels and lids together.
4 Options Available Below
Product Details & Documents
Stainless steel, rust-proof.
- Simple assembly and disassembly
- With three holding segments
- Quick release
For clamping reaction vessels and lids together.