Transfection boosters, GeneBlaster™
Supplier: OZ BIOSCIENCES
Certificates
About this item
GeneBlaster™ transfection boosters are an innovative and efficient solution to improving gene expression levels. GeneBlaster™ kits are a set of chemicals designed to get higher and longer transgene expression levels.
- Higher gene expression in many cells
- Convenient for a large panel of adherent and suspension cells
- Prolong in vitro gene expression
- Successful with all genetic vectors
- Simple, rapid, and easy to use
Store all GeneBlaster™ kits at +4 °C, except GeneBlaster™ Emerald at –20 °C.
Delivery: Shipped at room temperature.
Specifications
- Cell specific:Neurons