Orders LinkContactUsLinkComponent
Rectangular cuvettes
Rectangular cuvettes
Supplier:   BIOCHROM
Rectangular cuvettes
Supplier:  BIOCHROM
Certificates
Rectangular cuvettes
7 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • Height
    Volume
    Path length
    Window material
    Availability
    Price
  • SCLI80-2002-54
    Rectangular cell with lid
    Rectangular cell with lid
    45 mm
    200 µl
    1 mm
    UV silica
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • SCLI80-2003-83
    Rectangular cell with lid
    Rectangular cell with lid
    45 mm
    200 µl
    1 mm
    Optical glass
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • SCLI80-2002-57
    Rectangular cell with lid
    Rectangular cell with lid
    45 mm
    1 ml
    5 mm
    UV silica
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • SCLI80-2002-70
    Rectangular cell with stopper
    Rectangular cell with stopper
    48 mm
    2 ml
    10 mm
    UV silica
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • SCLI80-2003-87
    Rectangular cell with lid
    Rectangular cell with lid
    45 mm
    2 ml
    10 mm
    Optical glass
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • SCLI80-2002-58
    Rectangular cell with lid
    Rectangular cell with lid
    45 mm
    2,5 ml
    10 mm
    UV silica
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • SCLI80-2002-63
    Rectangular cell with lid
    Rectangular cell with lid
    45 mm
    10 ml
    50 mm
    UV silica
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST