Order Entry
Austria
ContactUsLinkComponent
Freund's complete adjuvant, CFAVax
Freund's complete adjuvant, CFAVax
Catalog # OZBICFA0010
Supplier:  OZ BIOSCIENCES
CAS Number:  
undefined
Freund's complete adjuvant, CFAVax
Catalog # OZBICFA0010
Supplier:  OZ BIOSCIENCES
CAS Number:  

Specifications

  • Pk:
    10 mL
  • Description:
    Freund's complete adjuvant, CFAVax, 10 ml
  • Seq:
    01

Specifications

About this item

Complete Freund’s Adjuvant (CFA) is a water-in-oil emulsion containing 1 mg per ml heat-killed Mycobacterium tuberculosis.

  • Suitable for the stimulation of Th1 response, initial immunisation and antibody production

It consists of a mixture of mineral oil and emulsifier in a ratio of 85% v/v oil and 15% v/v emulsifier. It is not a pre-formed emulsion and thus it must be mixed with an equal volume of aqueous solution of antigen and subsequently emulsified prior to use. The emulsifier is mannide monooleate, an ester consisting of mannitol as the hydrophilic residue and oleic acid (unsaturated C18 fatty acid) as the hydrophobic part.

Store at +4 °C

Delivery: Shipped at room temperature.
Caution: Intended for research use only.