Order Entry
Austria
ContactUsLinkComponent
Sulphuric acid 95.0-98.0% (by acidimetry), AR Select® ACS, Macron Fine Chemicals™
Corrosive
Irritant
Health
Sulphuric acid 95.0-98.0% (by acidimetry), AR Select® ACS, Macron Fine Chemicals™
Catalog # 5557-14
CAS Number:  
undefined
Corrosive
Irritant
Health
Sulphuric acid 95.0-98.0% (by acidimetry), AR Select® ACS, Macron Fine Chemicals™
Catalog # 5557-14
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, clear, Mall
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    95.0 - 98.0%
  • For Laboratory,Research,or Manufacturing Use:
    ≤0.5 ppm
  • ACS - Assay (H₂SO₄) (by acidimetry):
    95.0 - 98.0 %
  • Appearance:
    Passes Test
  • ACS - Ammonium (NH₄):
    ≤ 0.5 ppm
  • ACS - Arsenic and Antimony (as As):
    ≤ 0.003 ppm
  • ACS - Chloride (Cl):
    ≤ 0.05 ppm
  • ACS - Color (APHA):
    ≤ 8
  • ACS - Heavy Metals (as Pb):
    ≤ 0.1 ppm
  • ACS - Nitrate (NO₃):
    ≤ 0.1 ppm
  • ACS - Residue after Ignition:
    ≤ 1 ppm
  • ACS - Substances Reducing Permanganate (as SO₂):
    ≤ 2 ppm
  • Determine by Flame Photometry & ICP - Aluminum (Al):
    ≤ 50.0 ppb
  • Determine by Flame Photometry & ICP - Barium (Ba):
    ≤ 10.0 ppb
  • Determine by Flame Photometry & ICP - Beryllium (Be):
    ≤ 50.0 ppb
  • Determine by Flame Photometry & ICP - Bismuth (Bi):
    ≤ 100.0 ppb
  • Determine by Flame Photometry & ICP - Boron (B):
    ≤ 10.0 ppb
  • Determine by Flame Photometry & ICP - Cadmium (Cd):
    ≤ 5.0 ppb
  • Determine by Flame Photometry & ICP - Calcium (Ca):
    ≤ 50.0 ppb
  • Determine by Flame Photometry & ICP - Chromium (Cr):
    ≤ 10.0 ppb
  • Determine by Flame Photometry & ICP - Cobalt (Co):
    ≤ 5.0 ppb
  • Determine by Flame Photometry & ICP - Copper (Cu):
    ≤ 5.0 ppb
  • Determine by Flame Photometry & ICP - Gallium (Ga):
    ≤ 100.0 ppb
  • Determine by Flame Photometry & ICP - Germanium (Ge):
    ≤ 100.0 ppb
  • Determine by Flame Photometry & ICP - Gold (Au):
    ≤ 100.0 ppb
  • Determine by Flame Photometry & ICP - Iron (Fe):
    ≤ 50.0 ppb
  • Determine by Flame Photometry & ICP - Lead (Pb):
    ≤ 1.0 ppb
  • Determine by Flame Photometry & ICP - Lithium (Li):
    ≤ 10.0 ppb
  • Determine by Flame Photometry & ICP - Magnesium (Mg):
    ≤ 50.0 ppb
  • Determine by Flame Photometry & ICP - Manganese (Mn):
    ≤ 5.0 ppb
  • Determine by Flame Photometry & ICP - Mercury (Hg):
    ≤ 1.0 ppb
  • Determine by Flame Photometry & ICP - Molybdenum (Mo):
    ≤ 10 ppb
  • Determine by Flame Photometry & ICP - Nickel (Ni):
    ≤ 5.0 ppb
  • Determine by Flame Photometry & ICP - Potassium (K):
    ≤ 50.0 ppb
  • Determine by Flame Photometry & ICP - Silicon (Si):
    ≤ 100.0 ppb
  • Determine by Flame Photometry & ICP - Silver (Ag):
    ≤ 10.0 ppb
  • Determine by Flame Photometry & ICP - Sodium (Na):
    ≤ 500.0 ppb
  • Determine by Flame Photometry & ICP - Strontium (Sr):
    ≤ 10.0 ppb
  • Determine by Flame Photometry & ICP - Thallium (Tl):
    ≤ 200.0 ppb
  • Determine by Flame Photometry & ICP - Tin (Sn):
    ≤ 50.0 ppb
  • Determine by Flame Photometry & ICP - Vanadium (V):
    ≤ 10 ppb
  • Determine by Flame Photometry & ICP - Zinc (Zn):
    ≤ 5.0 ppb
  • Determine by Flame Photometry & ICP - Zirconium (Zr):
    ≤ 10 ppb

Specification Test Results

About this item

 :
 :98.08 g/mol
Merck Index:14,08974
Boiling Pt:∼335 °C (1013 hPa)
Melting Pt:3 °C
Density:1.84 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-639-5
UN:1830
ADR:8,II
 :Management of Change (MOC) category = N
 :GHS05
 :GHS07
 :GHS08
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589