Express Eingabe
Austria
ContactUsLinkComponent
Ammoniaklösung strong NF, ACS, J.T.Baker®
Corrosive
Irritant
Environment
Ammoniaklösung strong NF, ACS, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Corrosive
Irritant
Environment
Ammoniaklösung strong NF, ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    Passes Test
  • GMP Manufactured Product:
    ≤0.02%
  • Meets N.F. Requirements:
    Passes Test
  • CAUTION: For Manufacturing, processing or repackaging:
    27.0 - 30.0%
  • Bulk Pharmaceutical Chemical:
    28.0 - 30.0%
  • NF - Identification:
    Passes Test
  • NF - Nonvolatile Residue:
    ≤0.02 %
  • NF - Readily Oxidizable Substances:
    Passes Test
  • NF - Assay (NH₃)(NF):
    27.0 - 30.0 %
  • ACS - Assay (as NH₃):
    28.0 - 30.0 %
  • ACS - Carbon Dioxide (CO₂):
    ≤0.002 %
  • ACS - Residue after Ignition:
    ≤0.002 %
  • ACS - Substances Reducing Permanganate:
    Passes Test
  • Appearance:
    Passes Test
  • Arsenic and Antimony (as As):
    ≤3 ppm
  • Chloride (Cl):
    ≤0.5 ppm
  • Trace Impurities - Heavy Metals:
    ≤0.5 ppm
  • Nitrate (NO₃):
    ≤2 ppm
  • Phosphate (PO₄):
    ≤2 ppm
  • Sulfate (SO₄):
    ≤2 ppm
  • Trace Impurities - Aluminum (Al):
    ≤0.300 ppm
  • Trace Impurities - Boron (B):
    ≤0.100 ppm
  • Trace Impurities - Calcium (Ca):
    ≤0.300 ppm
  • Trace Impurities - Chromium (Cr):
    ≤0.200 ppm
  • Trace Impurities - Copper (Cu):
    ≤0.10 ppm
  • Trace Impurities - Gold (Au):
    ≤0.300 ppm
  • Trace Impurities - Iron (Fe):
    ≤0.2 ppm
  • Trace Impurities - Lead (Pb):
    ≤0.200 ppm
  • Trace Impurities - Magnesium (Mg):
    ≤0.300 ppm
  • Trace Impurities - Manganese (Mn):
    ≤0.200 ppm
  • Trace Impurities - Nickel (Ni):
    ≤0.1 ppm
  • Trace Impurities - Potassium (K):
    ≤0.300 ppm
  • Trace Impurities - Sodium (Na):
    ≤2.000 ppm
  • Trace Impurities - Tin (Sn):
    ≤0.300 ppm
  • Trace Impurities - Titanium (Ti):
    ≤0.300 ppm
  • Trace Impurities - Zinc (Zn):
    ≤0.300 ppm

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together