Orders LinkContactUsLinkComponent
J.T.Baker®, Liquid Level Indicators
J.T.Baker®, Liquid Level Indicators
Supplier:   Avantor
J.T.Baker®, Liquid Level Indicators
Supplier:  Avantor
Certificates
J.T.Baker®, Liquid Level Indicators
3 Options Available Below
Product Details & Documents
These level indicators provide the digital readout of the approximate volume of solvent remaining in the CYCLE-TAINER system.
  • Signal can also be used to drive switching circuits
Product Family Options
Product Information
  • Availability
    Price
  • 9803-03
    CYCLE-TAINER™ LCD liquid level volume display, for 52 L model C system
    CYCLE-TAINER™ LCD liquid level volume display, for 52 L model C system
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • 9803-01
    CYCLE-TAINER™ LCD liquid level volume display, for 52 L model C system
    CYCLE-TAINER™ LCD liquid level volume display, for 52 L model C system
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • 9803-00
    CYCLE-TAINER™ LCD liquid level volume display, for 215 L model C system
    CYCLE-TAINER™ LCD liquid level volume display, for 215 L model C system
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST