Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥98.0% (dry basis)
:77-86-1
:
:NH₂C(CH₂OH)₃
:MFCD00004679
:121.14 g/mol
:14,09772
:219…220 °C (10 mmHg)
:171…172 °C
:Ambient
:201-064-4
:GHS07
- Assay (Dry Basis):≥98.0%
- Identification (FTIR):Conforms
- Loss on Drying:≤1.5%
- pH (5%, H₂O):10.0 - 11.5
Frequently Bought Together