Order Entry
Austria
ContactUsLinkComponent
L-Arginine hydrochloride 98.5-101.5%
L-Arginine hydrochloride 98.5-101.5%
Certificates
L-Arginine hydrochloride 98.5-101.5%
Certificates

Specification Test Results

  • Assay (unspecified):
    98.5 - 101.5%
  • Impurity Content, Organic Volatile Impurities:
    Meets requirements
  • Impurity Content, Sulphate (SO₄):
    ≤0.03%
  • Loss on Drying:
    ≤0.20%
  • Optical Rotation:
    +21.4 - 23.6° (c=8 in 6 N HCl)
  • Identification (FTIR):
    Conforms
  • Chloride Content:
    16.5 - 17.1%
  • Residue on Ignition:
    ≤0.10%
  • Thin Layer Chromatography:
    Homogeneous
  • Heavy Metals (Pb):
    ≤20 ppm

Specification Test Results

Related Information

Frequently Bought Together