ContactUsLinkComponent
Empty disposable sample load cartridges, RediSep®
Empty disposable sample load cartridges, RediSep®
Catalog # HPLCAS-69-3873-235
Empty disposable sample load cartridges, RediSep®
Catalog # HPLCAS-69-3873-235
Specifications
  • Column Format:
    Flash cartridge
  • Bed weight:
    5 g
  • Pk:
    30
Specifications
About this item
RediSep® empty disposable sample load cartridges improve the resolution of the compound and eliminate reaction byproducts when compared to liquid injection techniques.
Prepare pre-filled solid load cartridges by pipetting the dissolved sample onto the top of the cartridge. Prepare empty solid load cartridges by filling the cartridge with a slurry mixture of the dissolved sample and supporting media. For optimal benefits, remove the solvent by vacuum before placing the cartridge on the purification system.