Order Entry
Austria
Orders LinkContactUsLinkComponent
 

VWR® Dressing Forceps, ABS

    
VWR® Dressing Forceps, ABS
    
 
VWR® Dressing Forceps, ABS
VWR® Dressing Forceps, ABS

 

 

Made of ABS (acrylonitrile butadiene styrene), purple.

  • Temperature resistance: –15 to +50 °C
  • Available in sterile or non sterile versions
  • EO sterilisation: Meets minimum requirement of SAL 10⁻⁶ (ISO 11137)

Shelf life: Expiration date is based on product and packaging shelf life, providing the product wrapper is not damaged.

 : Meets the requirements of BSE/TSE. Manufactured under the ISO 13485 standard.

 

 

  • Catalog No:
  • 232-0218
  • 232-0219
  • 232-0221
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • ISO 14001 (Certificate Registration No.131120)
  • ISO 14001 (Certificate Registration No.131120)
  • ISO 14001 (Certificate Registration No.131120)

 

Product Family Options

 

  • Type
    Colour
    Length
    Thickness
    Width
    Packed
     
     
  • Non sterile
    Purple
    110 mm
    1,5 mm
    9,9 mm
    100 per bag
       
       
    1 * 100 ST
    1 * 100 ST - 64,50 €
    64,50 €
    1 * 100 ST
       64,50 €

     
      Low Manufacturing Impact
     
    Specifications
    Cat. No.
    232-0218
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (Certificate Registration No.131120)
      Low Manufacturing Impact
  • Sterile
    Purple
    110 mm
    1,5 mm
    9,9 mm
    Individually packed
       
       
    1 * 100 ST
    1 * 100 ST - 80,70 €
    80,70 €
    1 * 100 ST
       80,70 €

     
      Low Manufacturing Impact
     
    Specifications
    Cat. No.
    232-0219
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (Certificate Registration No.131120)
      Low Manufacturing Impact
  • Sterile
    Purple
    130 mm
    1,6 mm
    13 mm
    Individually packed
       
       
    1 * 100 ST
    1 * 100 ST - 88,80 €
    88,80 €
    1 * 100 ST
       88,80 €

     
      Low Manufacturing Impact
     
    Specifications
    Cat. No.
    232-0221
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (Certificate Registration No.131120)
      Low Manufacturing Impact