Order Entry
Austria
ContactUsLinkComponent
Potassium hydroxide 0.1 mol/l (0.1N) traceable to NIST Standard Reference Material (SRM)
Potassium hydroxide 0.1 mol/l (0.1N) traceable to NIST Standard Reference Material (SRM)
Catalog # APPC181521.1211
Supplier:  PanReac AppliChem
CAS Number:  
Potassium hydroxide 0.1 mol/l (0.1N) traceable to NIST Standard Reference Material (SRM)
Catalog # APPC181521.1211
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pk:
    1.000 mL
  • Seq:
    2865

Specifications

Specification Test Results

  • Titer at 20 °C:
    0.999 - 1.001
  • Traceability:
    NIST

Specification Test Results

About this item

 :
 :56.11 g/mol
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553