Order Entry
Austria
ContactUsLinkComponent
NMR sample tubes, ECONOMY, 3 mm, SP Wilmad-LabGlass
NMR sample tubes, ECONOMY, 3 mm, SP Wilmad-LabGlass
  WILMWG-3000-7-50
NMR sample tubes, ECONOMY, 3 mm, SP Wilmad-LabGlass
  WILMWG-3000-7-50

 

  • Type:
    With cap
  • Length:
    177,8 mm
  • Seq:
    03
  • Pk:
    50

 

 

High-throughput NMR tubes are made of borosilicate glass Type I, class A.

  • Designed for routine use in most low to mid field NMR spectrometers
  • One of the best OD tolerances in the industry
  • Guaranteed to fit tightly in spinner turbine
  • 100% inspected for surface defects and physical dimension to ensure the success of your experiments