Order Entry
Austria
ContactUsLinkComponent
Racks for containers, with lid
Racks for containers, with lid
Catalog # DELTNT-250
Supplier:  DELTALAB
Racks for containers, with lid
Catalog # DELTNT-250
Supplier:  DELTALAB

Specifications

  • Description:
    Rack with 12 holes 63 mm diameter
  • Pk:
    1

Specifications

About this item

Made of stainless steel wire with lid.

  • Hole of 63 mm
  • Able to carry the containers of up to 58 ml Ø

Mainly designed for carrying containers, as they stand firmly in the rack thanks to the lid.