ContactUsLinkComponent
Disposable syringes, Norm-Ject®
Disposable syringes, Norm-Ject®
Catalog # MEGRHSW4200.090D0
Supplier:  MEGRO
undefined
Disposable syringes, Norm-Ject®
Catalog # MEGRHSW4200.090D0
Supplier:  MEGRO
Specifications
  • Capacity:
    20 ml
  • Description:
    Syringe, 2-parts
  • Seq:
    02
  • Environmentally Preferable:
  • Pk:
    1.000
Specifications
About this item
Standard-disposable syringe, easy working and absolutely leak-proof, clear as crystal cylinder, scale is easy to read, with a second braking ring on the bottom of the cylinder.
  • Piston is made of polyethylene
  • Cylinder is made of polypropylene
Second braking ring on the bottom of the cylinder. thereby the resistance will be increased to separate the piston and the cylinder, free of latex and silicone oil.