ContactUsLinkComponent
1536-well microplates, cyclo olefin
1536-well microplates, cyclo olefin
Catalog # GREI792870-906
Supplier:  Greiner Bio-One
   Low Manufacturing Impact
1536-well microplates, cyclo olefin
Catalog # GREI792870-906
Supplier:  Greiner Bio-One
   Low Manufacturing Impact
Specifications
  • Form:
    F-Bottom
  • Packed:
    15/ bag
  • Recommended working volume:
    3 - 10 µl
  • Sterile:
    -
  • Colour:
    Clear
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO 14001 (241002); ISO 50001 (241002)
  • Pk:
    60
  • Well volume:
    12,6 µl
Specifications
About this item
Clear, 1536-well cycloolefin storage plates (with optimised geometry for low evaporation) for acoustic liquid handling.
  • Free from DNase, RNase, and human DNA
  • Groove around the plate edge matches cycloolefin plate lid (not included) - designed to reduce evaporation and minimise edge effects
Acoustic liquid dispensing offers the advantage of pipetting small volumes very precisely and with minimal disturbance without the risk of cross-contamination. The process involves the transferral of DMSO-containing solutions in the nanoliter to picoliter range from a starting plate into a target plate by means of ultrasound.
Certifications: Manufactured under DIN ISO 9001 guidelines.
Ordering information: Plates are supplied without lids. Accessory lid is sold separately.