Order Entry
Austria
ContactUsLinkComponent
TraceCERT®, Supelco® Bromide Standard for Ion Chromatography (IC)
TraceCERT®, Supelco® Bromide Standard for Ion Chromatography (IC)
Catalog # SUPL43147-100ML
Supplier:  Merck
CAS Number:  
TraceCERT®, Supelco® Bromide Standard for Ion Chromatography (IC)
Catalog # SUPL43147-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pk:
    100 mL
  • Description:
    Bromide
  • M-Clarity quality level:
    MQ300

Specifications

About this item

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34). This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST or BAM.