Order Entry
Austria
Orders LinkContactUsLinkComponent
PBS tablets

PBS tablets

PBS tablets, 7,4 (25 °C), 100
Catalog # A9162.0100
Supplier:   PanReac AppliChem
PBS tablets
Catalog # A9162.0100
Supplier:  PanReac AppliChem
CAS Number:  
PBS tablets
PBS tablets

PBS tablets, 7,4 (25 °C), 100

CAS Number:  
Price
162,00 €
Log in for availability
1 * 100 ST
Product Details & Documents

Tablets for preparation of 100 ml phosphate buffered saline (PBS).

  • Ready to use
  • Autoclaveable

Deposit one tablet in flask or beaker placed on a magnetic stirrer. Add deionised water to 50% of the final volume at room temperature. Stir for a few minutes. Adjust the water to the final volume, stir until full dissolution. Autoclave or filter sterilise through a 0,22 μm filter into a sterile flask or bottle.

Application includes tissue culture or cell culture, sample dilution or protein dilution, immunoassays or immuno-histochemistry and microbiology.

Documents

Specifications

Specifications

  • Pk:
    100
  • Description:
    PBS tablets
  • pH value:
    7,4 (25 °C)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.