


N,N-Dimethylformamide ≥99.8% (GC, on anhydrous basis), PESTINORM® for pesticide residue analysis
:68-12-2
:
:HCON(CH₃)₂
:MFCD00003284
:73.09 g/mol
:–60.5 °C
:Ambient
:200-679-5
:2265
:3,III
:Filtered through a 0.2 µm filter, packaged under nitrogen.
:GHS02
:GHS08
:GHS07
- Appearance:Clear, colourless liquid
- Assay (GC, on anhydrous basis):min. 99.8 %
- Identity (IR):Comply
- Residue after evaporation:max. 0.0003 % w/w
- Water (KF):max. 0.05 % w/w
- Color (APHA, Pt-Co):max. 10
- GC/ECD any Pesticide (as Lindane).:max. 5 pg/ml
Frequently Bought Together