Order Entry
Austria
ContactUsLinkComponent
Ethanol absolute ≥99.8%, PESTINORM® for pesticide residue analysis
Flammable
Irritant
Ethanol absolute ≥99.8%, PESTINORM® for pesticide residue analysis
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates
undefined
Flammable
Irritant
Ethanol absolute ≥99.8%, PESTINORM® for pesticide residue analysis
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates
Limited Time Sale
Select items below are currently on sale

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion

Specifications

Specification Test Results

  • Appearance:
    Clear colorless liquid
  • Assay (GC, on anhydrous basis):
    min. 99.8 %
  • Identity (IR):
    Comply
  • Residue after evaporation:
    max. 0.0003 % w/w
  • Water (KF):
    max. 0.05 % w/w
  • GC/ECD any Pesticide (as Lindane):
    max. 5 pg/ml

Specification Test Results

Related Information

Frequently Bought Together