Order Entry
Austria
ContactUsLinkComponent
Sulphuric acid concentrate for dilution to 0.005 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
Sulphuric acid concentrate for dilution to 0.005 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
Catalog # 84102.600
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sulphuric acid concentrate for dilution to 0.005 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
Catalog # 84102.600
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic ampoule
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

Specification Test Results

  • Titer (N):
    0.0099-0.0101

Specification Test Results

About this item

 :
 :98.08 g/mol
 :Dose for preparation of 1 litre of solution.
Storage Temperature:Ambient
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589