ESD safety ankle boots, lace-up, Sander
Supplier: ELTEN
Certificates
About this item
100% metal- and leather-free, lightweight ESD safety ankle boot made from hydrophobised microfibre and hydrophobised CORDURA® (tear and abrasion resistant, dirt and water repellent), a padded tongue plus a slip resistant, dual-density TPU/PU sole (Trainers).
- Climate-regulating, breathable textile lining
- Replaceable insole; orthopaedic insoles possible
- Full length fleece inlay sole (ESD Pro), black
- Composite toe protection cap (200 J)
Ideal for use in areas where exposure to moisture is expected.
Colour: Black/red
Certifications: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S2, SRC
Specifications
- Catalog No:
- ELTN76872/38
- ELTN76872/41
- ELTN76872/48
- ELTN76872/35
- ELTN76872/36
- ELTN76872/46
- ELTN76872/47
- ELTN76872/44
- ELTN76872/45
- ELTN76872/42
- ELTN76872/43
- ELTN76872/37
- ELTN76872/39
- ELTN76872/40
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001